Learn More
Invitrogen™ TRA-1-60 (Podocalyxin) Monoclonal Antibody (TRA-1-60), PerCP-eFluor™ 710, eBioscience™

Description
The TRA-1-60 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-60 antibody is lost upon cell differentiation. Contrary to early reports that the TRA-1-60 epitope can be destroyed by neuraminidase digestion, new strudies have shown that TRA-1-60 recognizes a neurominidase resistent antigen. The TRA-1-60 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer. Applications Reported: This TRA-1-60 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TRA-1-60 antibody has been tested by flow cytometric analysis of the 2102Ep cell line. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal pe...
Specifications
Specifications
| Antigen | TRA-1-60 (Podocalyxin) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TRA-1-60 |
| Concentration | 0.2 mg/mL |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene Alias | TRA160; tra60 |
| Host Species | Mouse |
| Purification Method | Affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.