Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-81 Mouse anti-Human, NL493, Clone: TRA-1-81, R&D Systems™

Catalog No. NLLC16581G
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NLLC16581G 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NLLC16581G Supplier R&D Systems Supplier No. NLLC16581G
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRA-1-81 Monoclonal antibody specifically detects TRA-1-81 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen TRA-1-81
Applications Immunocytochemistry
Classification Monoclonal
Clone TRA-1-81
Conjugate NorthernLights 493 (NL493)
Dilution Immunocytochemistry
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias GCTM, gp200, MGC138240, PC, PCLP, POD XL, PODXL, TRA181
Gene Symbols PODXL
Host Species Mouse
Quantity 0.5 mL
Primary or Secondary Primary
Target Species Human
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date. Protect from light.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.