Learn More
Sartorius TRACE MEMB DIALYS GLC/LAC 5PK
Supplier:  Sartorius BPT0024
Encompass
Trace Membranes Dialysis for glucose/lactate with higher measuring rangefor the dialysis probe comes in a package of 5 pieces. The membranes are part of the BioPAT Trace and Multi Trace System. In Addition the membranes are ready to use can easily be installed in the dialysis probe and replaced after every cultivation.
Catalog No. NC2398123
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov