Learn More
Fisherbrand™ Traceable™ 7600 Door Sensor
Catalog No. 01257637 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Traceable 7600 Smart Wi-Fi Data Loggers
Catalog No. For Use With (Equipment)
01-257-637 Traceable 7600 Smart Wi-Fi Data Loggers
1 options
Catalog No. 01-257-637
Add to Cart
Add to Cart
Open-door sensors prevent accidental temperature excursions
  • Each data logger can support up to two open-door sensors
  • Sensors will alert if a door is accidentally left open
Specifications
Type Sensor
Warranty 1 year
For Use With (Equipment) Traceable 7600 Smart Wi-Fi Data Loggers