Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Excursion-Trac™ Datalogging Thermometer
SCI_ED

Catalog No. p-6970036 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
1 Bottle Probe
1 Probe, 5/16 in. dia. x 3/4 in. long
1 Stainless-Steel Probe
2 Bottle Probes
2 Probes, 5/16 in. dia. x 3/4 in. long
2 Stainless-Steel Probes
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S2-510-3 1 Bottle Probe
S2-510-4 2 Bottle Probes
S2-510-6 2 Probes, 5/16 in. dia. x 3/4 in. long
S2-510-8 1 Stainless-Steel Probe
S2-510-9 2 Stainless-Steel Probes
S2-510-5 1 Probe, 5/16 in. dia. x 3/4 in. long
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S2-510-3
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Stand, Velcro™, magnetic strips, wall mount, AC power adapter, batteries, and an ISO 17025 Traceable™ Certificate

Monitor temperatures in refrigerators, freezers, water baths, heating blocks and incubators with this datalogging thermometer. Traceable™ Excursion-Trac™ Datalogging Thermometers have a temperature range of -50 to 70°C and resolution of 0.01 and maintain an accuracy of ±0.25°C across the entire temperature range.

Datalogging thermometer with user-defined time intervals

  • Stores up to ten unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Datalogging Capacity: 525,600 temperature observations
  • Weight: 5 oz.

Hi/Lo Alarms and Time/Date Stamps

  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display

USB Data Transferring

  • New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location

Data-Logging Features

  • Monitor readings overnight, on weekends, or for any time period with rolling data log
  • User-defined data-logging interval of one observation/minute to one observation/24 hours
  • Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals

Bottle Probe Features (Cat. Nos. 15-081-125, -126, -128, 129 do not come with bottle probes)

  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples

Note: Batteries are designed for power back-up and will last for 7 days. This unit comes standard with an AC power adapter, which is recommended for standard use.

Order Info

Replacement battery: Cat. No. 1111

Certifications

Supplied with an ISO 17025 Traceable™ Certificate

Specifications

Type Datalogger
Display Type LCD
Data Memory 525,600 Readings
Calibration Not Calibrated
Alarm Types LED Indicator, Audible, Flashing LCD Segment
Software Type TraceableLIVE
Description 1 Bottle Probe
Includes 1 Bottle Probe
Certifications/Compliance ISO 17025, A2LA, NIST
Interface USB
Sensor Types Temperature-Buffered Bottle Sensor
Accuracy (Temperature) ±0.25°C
Temperature Resolution 0.01°
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50.00°C to 70.00°C
Length (English) Cable 10 ft.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.