Learn More
Thomas Scientific Traceabler Flash LED Alert Dual Channel Timer
SDP
Supplier:  Thomas Scientific 1222W24
Encompass_Preferred
Flashing LED indicator 2-seperate timing channels Automatic programmable memory Two channel timing range is 99 hours, 59 minutes, 59 seconds Resolution is 1 second; accuracy is 0.01% Size is 3-1/2" x 3" x 5/8"; weight is 3 ounces
Catalog No. NC0783417