Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Memory-Loc™ Datalogging Thermometer

Catalog No. 15079620 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
1 Bottle
1 Probe, 5/16 in. dia. x 3/4 in. long
1 Stainless-steel Probe
1 Vaccine Bottle Probe
2 Bottles
2 Probes, 5/16 in. dia. x 3/4 in. long
2 Stainless-steel Probes
2 Vaccine Bottle Probes
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15-079-620 2 Vaccine Bottle Probes
15-081-130 1 Bottle
15-081-136 2 Stainless-steel Probes
15-081-132 1 Probe, 5/16 in. dia. x 3/4 in. long
15-081-131 2 Bottles
15-081-135 1 Stainless-steel Probe
15-081-133 2 Probes, 5/16 in. dia. x 3/4 in. long
15-079-619 1 Vaccine Bottle Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15-079-620
Add to Cart
Add to Cart

Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.

  • Data-logging thermometer with fixed one-minute timing intervals
  • Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
  • Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
  • Stores up to ten unique alarm events
  • Capable of storing over 1 million temperature observations
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, low memory, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Weight: 5 oz.

Hi/Lo Alarms and Time/Date Stamps

  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display

USB Data Transferring

  • All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
  • Raw data on unit cannot be overwritten or modified
  • Indicator light will appear when 95% of memory is full

Data-Logging Features

  • Can be used in environments requiring 21 CFR 11 compliance
  • Thermometer logs two years of observations with a one-minute logging interval
  • Device will stop recording when storage is full and will not overwrite recorded data
  • Data is unable to be deleted from memory once stored

Bottle Probe Features

  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples
  • Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program

Order Info

Replacement battery: Cat. No. 1111

Specifications

Type Datalogger
Display Type Digital
Alarm Types Audible, Visual
Software Type TraceableLIVE
Description 2 Vaccine Bottle Probes
Includes 2 Vaccine Bottle Probes
Certifications/Compliance NIST, ISO/IEC 17025, A2LA, CDC
Accuracy (Temperature) ±0.25°C
Temperature Resolution 0.01°
Temperature Range (Metric) -50°C to +70°C
Operating Temperature (English) -58°F to +158°F
Operating Temperature (Metric) -72.4°F to +316.4°F
Weight (English) 5 oz.
Weight (Metric) 141.7 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.