Learn More
Traceable™ One-Shot™ Conductivity Calibration Standards Certified Reference Material Producer (CRM), Fisherbrand™
Shop All Fisherbrand ProductsDescription
Ideal for lab or field applications
Single-use conductivity standards calibrate all conductivity meters and probes for maximum accuracy. Eliminate concern about external container contamination. Calibration is made in the standard vial.
Calibration within container — zero contamination
- Traceable to NIST (National Institute of Standards and Technology)
- Accuracy: certificate stated uncertainty or ±0.25 microsiemen or ±0.25%
- 100% compatible with all conductivity meters
- A2LA accreditations ISO 34 and ISO 17025 plus ISO 9001
- Includes an individual temperature chart and step-by-step calibration instructions
- Units labeled (micromhos/microsiemens, ohms, parts per million/TDS)
- Single use Eliminates external contamination concerns Each standard contains 100mL
Assortment (09-328-20) contains one each of 09-328-6, -7, -8, -9, -10, and -12
Order Info
Each bottle contains 100mL
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | 6/Pk. |
| Conductivity | 10, 100, 1000, 1413, 10,000, 100,000 μ℧ |
| Technique | Calibration of Conductivity Instrument |
| Color | Clear |
| pH | 7 to 9 |
| For Use With (Equipment) | Conductivity Meter |
| Solubility | Soluble in water |
| Solution Type | Calibration Solution |
| Type | Conductivity Standard |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.