Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer
SCI_ED

Catalog No. S14226 Shop All Fisherbrand Products
Change view
Click to view available options
Probe Type:
1 Bottle Probe
1 Bottle and 1 Bullet Probe
1 x 5 mL Vaccine Bottle Probe
2 x 5 mL Vaccine Bottle Probe
One Bullet Probe
Two Bottle Probes
Two Bullet Probes
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Probe Type
S14226 1 x 5 mL Vaccine Bottle Probe
S14225 1 Bottle and 1 Bullet Probe
S14227 2 x 5 mL Vaccine Bottle Probe
S14222 Two Bottle Probes
S14224 Two Bullet Probes
S14221 1 Bottle Probe
S14223 One Bullet Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. S14226
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.

  • Triple display simultaneously shows min., max. and current temperatures for a specified probe
  • Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
  • Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
  • °C/°F switchable
  • Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
  • Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
  • Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
  • Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Range: -50° to +70°C (-58° to +158°F)
  • Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
  • Resolution: 0.1°

Certifications

Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.

Specifications

Bottle Fill Nontoxic Glycol
Bottle Size 2.5 x 6.3 cm (1 x 2.5 in.)
Certifications/Compliance NIST Traceable
Dimensions (L x W x H) 7 x 10.8 x 2 cm (2.75 x 4.25 x 0.75 in.)
For Use With (Application) Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators
Includes Traceable™ Certificate, flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
Length (Metric) 10.8 cm
Material ABS Plastic
Probe Type 1 x 5 mL Vaccine Bottle Probe
Resolution 0.1°
Weight (English) 4 oz.
Weight (Metric) 113.4 g
Product Type Refrigerator/Freezer Thermometer
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Refrigerators/Freezers
Accuracy ±0.5°C (-30°C to +50°C), ±1°C otherwise
Display Digital
Width (Metric) 6.99 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.