Learn More
Thomas Scientific TRACEABLE TEM DATA LOGGER AMB
SDP
Supplier:  Thomas Scientific 1157G90
Encompass
Thomas® Traceable® Temperature Bluetooth Data Logger compatible with TraceableGO™ App and TraceableLIVE® Cloud Service ; Ambient Sensor, -20.00 to 70.00°C
Catalog No. NC2877711