Learn More
Thomas Scientific TRACEABLE ULTRA ROBO THERM
SDP
Supplier:  Thomas Scientific 1235D17
Encompass
Traceable Robo-Thermometer Ultra, Accuracy ±0.4°C; Traceable to NIST for accuracy; Long stem allows the user to make measurements while stirring solutions. It is ideal for use in deep vessels.
Catalog No. NC1705745