Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ TraceableGO™ Bluetooth Datalogging Thermometer

Catalog No. 15079678 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-20°C to +70°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
15-079-678 -20°C to +70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-079-678
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceableGO™ Bluetooth Datalogger fills in the unknowns between destinations.

  • Fills in the unknowns between destinations
  • Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times
  • Simple, single button use
  • Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
  • Compatible with TraceableGO or TraceableLIVE Mobile Apps:
    • Once the trip has ended,users may export the data
    • App immediately identifies if the contents experienced conditions outside user-defined parameters
    • View if an alarm was triggered
    • Generate an instant data report
    • Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
    • Logging duration is 227 days (7.5 months)
  • TraceableGO™ Bluetooth devices are pre-programmed with alarm values

Certifications

ISO 17025, A2LA, NIST

Warranty and Services

1 year from date of purchase

Specifications

Type Temperature Datalogger
Display Type LCD
Data Memory 90 Alarm Events, 65,536 Temperature Readings
Alarm Types High/Low
Software Type TraceableLIVE
Battery Type 2 x AAA
Includes Batteries, wall mount, Traceable™ Certificate
Certifications/Compliance ISO 17025, A2LA, NIST
For Use With (Application) Temperature monitoring
Accuracy ±0.4°C between -10 and 70°C, otherwise ±0.5°C
Response Time 5 sec.
Temperature Resolution 0.1°
Temperature Range (English) -4°F to +158°F
Temperature Range (Metric) -20°C to +70°C
Height (English) 2 in.
Height (Metric) 5.1 cm
Length (English) 3.5 in.
Length (Metric) 8.9 cm
Width (English) 0.8 in.
Width (Metric) 2.0 cm
Weight (English) 0.37 lb.
Weight (Metric) 0.17 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.