Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ TraceableGO™ Temperature Data Logger with Bluetooth™, Calibration and Bottle Probe

Catalog No. 15079701 Shop All Fisherbrand Products
Change view
Click to view available options
Data Memory:
64,000 Readings, 7.5 Months, 5 min. Interval
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Data Memory
15-079-701 64,000 Readings, 7.5 Months, 5 min. Interval
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-079-701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easily access data from any location with Bluetooth™ data loggers.

  • Critical Information (Temperature, Trip Duration, MIN/MAX, Battery Level and Alarms) displayed on LCD at all times Simple, single button use
  • Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location
  • Compatible with TraceableGO or TraceableLIVE Mobile Apps:
    • Once the trip has ended, users may export the data
    • App immediately identifies if the contents experienced conditions outside user-defined parameters
    • View if an alarm was triggered
    • Generate an instant data report
    • Device stores up to 90 alarm events, records readings once every 5 minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours
    • Logging duration is 227 days (7.5 months)
  • TraceableGO™ Bluetooth devices are pre-programmed with alarm values

Specifications

Type Temperature Datalogger
Display Type Multi-line LCD
Data Memory 64,000 Readings, 7.5 Months, 5 min. Interval
Alarm Types High/Low
Software Type TraceableLIVE
Battery Type 2 AAA
Description Bluetooth datalogging thermometer with bottle probe
Includes Batteries, wall mount, Traceable™ Certificate
Channels 1
Certifications/Compliance ISO 17025, NIST
Interface Bluetooth
For Use With (Application) Temperature monitoring
Accuracy (Temperature) ±0.3°C
Temperature Resolution 0.1°
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Dimensions (L x W x H) 8.9 x 2 x 5.1 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Length (English) 8.9 cm
Length (Metric) 3.5 in.
Width (English) 0.8 in.
Width (Metric) 2 cm
Weight (English) 6 oz.
Weight (Metric) 0.17 kg
Show More Show Less

Option to subscribe to cloud-based TraceableLIVE™ service to upload data to cloud.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.