Learn More
Fisherbrand™ TraceableLIVE™ Ultra-Low Temperature WiFi Datalogging Thermometer with Remote Notification
SCI_ED
Catalog No. S24974 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
1 Stainless-steel 316 Probe, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
2 Stainless-steel 316 Probes, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S2-497-4 1 Stainless-steel 316 Probe, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
S2-497-5 2 Stainless-steel 316 Probes, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S2-497-4
Add to Cart
Add to Cart
Requires
TracebleLIVE requires subscription
Now with FREE TraceableLIVE subscription plan (for one user)! A simple, efficient, and reliable way to stay connected to your critical environments with smartphone, tablet or personal computer.
Use the TraceableLIVE™ app†, or visit Traceablelive.com† on any browser to view your current temperatures, control your alarm parameters, view your datalogging history, view and run reports or give other team members access to the data.
  • Remotely monitor temperature and more by connecting to existing WiFi
  • Alerts: temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface that requires no local software; web, Android, Apple IOS data visibility
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive Mobile Push/E-Mail/Text notifications
  • Unlimited scalability — use TraceableLIVE™ in individual labs or an entire network of facilities
  • Multiple point temperature calibration traceable to NIST
†Requires TraceableLIVE subscription
TraceableLIVE Subscription Cost through traceablelive.com:
  • Monthly Payment Plan: $8.99 per device for 1-9 devices or $7.20 per device for 10+ devices
  • Yearly Payment Plan: $100 per device for 1-9 devices or $80 per device for 10+ devices
  • Demo TraceableLive software at traceablelive.com
Specifications
Type Temperature Datalogger
Display Type Digital
Alarm Types Audible, Visual
Software Type TraceableLIVE
Includes 1 Stainless-steel 316 Probe, Quick Start Guide, batteries, magnets, Velcro, wall mount, and Traceable™ Certificate
Certifications/Compliance Traceable Certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA
For Use With (Application) Remotely monitor temperature
Accuracy (Temperature) ±0.1°C
Temperature Resolution 0.01°
Temperature Range (Metric) -90.00°C to +105.00°C