Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ TraceableOne™ 10 Pack Single-User Datalogger

Catalog No. 15079689 Shop All Fisherbrand Products
Change view
Click to view available options
Description:
Preprogrammed -20°C to -15°C
Preprogrammed 2°C to 8°C
Alarm Types:
Preprogrammed -20°C to -15°C
Preprogrammed 2°C to 8°C
Data Memory:
16,000 Readings, 10 Day, 1 min. Interval
16,000 Readings, 60 Day, 6 min. Interval
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Alarm Types Data Memory
15-079-689 Preprogrammed 2°C to 8°C Preprogrammed 2°C to 8°C 16,000 Readings, 10 Day, 1 min. Interval
15-079-694 Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C 16,000 Readings, 60 Day, 6 min. Interval
15-079-690 Preprogrammed -20°C to -15°C Preprogrammed -20°C to -15°C 16,000 Readings, 10 Day, 1 min. Interval
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 15-079-689

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 10 Pack Single-User Datalogger allows for monitoring materials every step of the way.

  • Logger generates PDF report
  • Download directly to PC
  • No Software required
  • Compact
  • Single-use
  • Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
  • Press start to begin recording crucial temperature data, 30 minute time delay before logging begins

Specifications

Type Datalogger
Data Memory 16,000 Readings, 10 Day, 1 min. Interval
Calibration Calibrated
Alarm Types Preprogrammed 2°C to 8°C
Software Type No software required, download data as PDF report
Description Preprogrammed 2°C to 8°C
Includes Batch-calibrated Traceable certificate
IP Rating IP67
For Use With (Application) Refrigerated transportation samples
Accuracy (Temperature) ±0.5°C from -20° to 40°C, otherwise ±1°C
Temperature Resolution 0.01°
Temperature Range (Metric) -30°C to +70°C
Length (Metric) 8.89 cm
Electrical Requirements Battery-Powered
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.