Learn More
TraceCERT™ 1,3,5-Trimethylbenzene Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 108-67-8 |
| Flash Point | 53°C |
| Synonym | Mesitylene solution |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 5,000 μg/mL in methanol |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.