Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ 2-Butanone Solution, 2,000 μg/mL in Methanol: water (9:1), MilliporeSigma™ Supelco™

Catalog No. 111020258 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0258 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0258 Supplier MilliporeSigma Supelco Supplier No. 48877

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

2-Butanone can play the role of an oxidant in the oppenauer oxidation of primary alcohol like 4-methoxybenzyl alcohol to the corresponding aldehyde, p-anisaldehyde.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
Ignition Point 960°F
CAS 78-93-3
Vapor Pressure 71 mmHg (20°C)
Vapor Density 2.49 (vs. air)
Density 0.805 g/mL (at 25°C)
Flash Point 9°C
Linear Formula C2H5 COCH3
MDL Number MFCD00011648
Melting Point -87°C
Boiling Point 80°C
Synonym 2-Butanone; Ethyl methyl ketone; MEK; Methyl ethyl ketone
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 72.11
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in methanol: water (9:1)
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.