Learn More
TraceCERT™ Diuron certified reference material, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 330-54-1 |
| Linear Formula | C9H10 Cl2N2 O |
| MDL Number | MFCD00018136 |
| Synonym | 3-(3,4-Dichlorophenyl)-1,1-dimethylurea |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 233.09 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.