Learn More
TraceCERT™ EPA 8270 Semivolatile Internal Standard Mix, 2,000 μg/mL, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL each component in methylene chloride |
| Quantity | 2 x 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | Acenaphthene-d10; Chrysene-d12; 1,4-Dichlorobenzene-d4; Naphthalene-d8; Perylene-d12; Phenanthrene-d10 |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.