Learn More
Eppendorf™ Trade-in Reference™ 2
Catalog No. 05414262 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
Catalog No. Volume (Metric)
05-414-262 0.1 to 2.5 μL
1 options
This item is not returnable. View return policy
Catalog No. 05-414-262 Supplier Eppendorf™ Supplier No. 2231302066
Add to Cart
Add to Cart
This item is not returnable. View return policy
Trade-in Reference™ 2 - Spring-loaded tip cone improves user-to-user reproducibility and ergonomics by lowering the tip attachment force.
  • Returned pipettes do not have to be in working condition and may be any manufacturers brand.
  • Pipettes must be free of contamination and returned together with the signed Decontamination Form (www.eppendorf.com/decontamination) to the following address:
    • In U.S.: Eppendorf Services, 151 Memorial Drive, Suite B, Shrewsbury, MA 01545
    • In Canada: Eppendorf Canada, 2810 Argentia Road, Unit #2, Mississauga, ONT L5N 8L2
    • If you have any questions, please contact Customer Support at :
    • U.S.: 800-645-3050
    • Canada: 800-263-8715
  • Spring-loaded tip cone improves user-to-user reproducibility and ergonomics by lowering the tip attachment force
  • The unique smooth surface and full autoclavability guarantee efficient decontamination - making it the ideal companion when working under sterile conditions
  • A temporary adjustment allows for an easy adjustment of your Reference 2 for the most accurate pipetting of different liquids or in changing external conditions without the need for a full calibration
  • Available as single-channel pipette in fixed or variable volume as well as 8- and 12-channel pipette
  • The Reference 2 multi-channel variants offer high flexibility incl. a spring-loaded tip cone which can be switched on/off optionally
TRUSTED_SUSTAINABILITY
Specifications
Color Dark Gray
Number of Channels 1
Product Line Reference 2
Type Pipette
Volume (Metric) 0.1 to 2.5 μL