Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL102 Medium Duty Synthetic Leather Mechanics Gloves

Catalog No. 19555037 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-037 Large
19-555-039 Small
19-555-038 Medium
19-555-041 X-Large
19-555-040 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-037 Supplier Pyramex Safety Products Supplier No. GL102L
This item is not returnable. View return policy

Gloves feature thicker embossed neoprene on the knuckles for protection, with touchscreen compatibility on two fingers and thumb.

  • Double layer spandex with mesh for comfort and breathability
  • Silicone palm patch for anti-slip and enhanced grip capabilities
  • Thicker embossed neoprene on knuckles for additional protection
  • Touch screen function on 2 fingers and thumb

Specifications

Back Material Mesh
Coating Material Silicone
Color Black
Hand Specific
Material Synthetic Leather, Spandex
Product Type Mechanic's Gloves
Size Large
Finger Style Full
For Use With (Application) Mechanics, Construction, Fabrication, and Assembly
Glove Cuff Style Hook and Loop
Series GL102
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.