Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TRAF3IP2 Antibody Blocking Peptide
SDP

Catalog No. NB398663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398663 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398663 Supplier Novus Biologicals™ Supplier No. NB10056740PEP
Add to Cart
Add to Cart

Specifications

Gene ID (Entrez) 10758
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56740
Gene Alias ACT1DKFZP586G0522, adapter protein CIKS, C6orf2, C6orf4chromosome 6 open reading frame 5, C6orf5DKFZp586G0522, C6orf6MGC3581, CIKSchromosome 6 open reading frame 2, Connection to IKK and SAPK/JNK, NFkB-activating protein ACT1, Nuclear factor NF-kappa-B activator 1, TRAF3 interacting protein 2, TRAF3-interacting protein 2
Gene Symbol TRAF3IP2
Quantity 0.1 mg
Research Category Signal Transduction
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.