Learn More
Invitrogen™ TRAF7 Polyclonal Antibody

Description
Suggested positive control: antigen standard for TRAF7 (transient overexpression lysate).
Specifications
Specifications
| Antigen | TRAF7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | TRAF7 |
| Gene Accession No. | Q6Q0C0, Q922B6 |
| Gene Alias | DKFZp586I021; E3 ubiquitin-protein ligase TRAF7; MGC7807; RFWD1; RGD1559653; ring finger and WD repeat domain 1; ring finger and WD repeat domain 1 isoform 1; RING finger and WD repeat-containing protein 1; RING finger protein 119; RING-type E3 ubiquitin transferase TRAF7; RNF119; TNF receptor associated factor 7; TNF receptor-associated factor 7; TNF receptor-associated factor 7, E3 ubiquitin protein ligase; traf7 |
| Gene Symbols | TRAF7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.