Learn More
Description
- Durable, .080 in. aluminum signs will not rust
- Engineer-Grade Prismatic Reflective sign face provides visibility in low-light conditions
- UV-resistant inks prevent fading from long-term outdoor use
- Clear UV Protective Overcoat
- Post mount holes
- Meets ASTM D 4956 Type I
Specifications
Specifications
| Certifications/Compliance | ASTM D 4956 Type I |
| Grade | Engineer |
| Material | Reflective Aluminum |
| Mounting Type | Corner Holes |
| Languages | English |
| Dimensions (L x W) | 18 x 24 in. |
| Displayed Text | EMERGENCY MEDICAL CARE |
| Color | White on Blue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.