Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGE™ PEEKsil™ Injection Loops
SDP

Catalog No. SG0650010 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
SG-0650010 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SG-0650010 Supplier SGE™ Supplier No. 0650010

missing translation for 'validMainframeMsg'

Compatible with most valves including Rheodyne™ and Valco™

  • Smooth, non-reactive internal bore reduces sample carry-over and sample rinse times
  • Reduced sample loss due to shear or adsorption
  • Maximum operating pressures: 3000psi for 10μL and 20μL loops
  • Supplied with PEEKsil tubing and two sets of Stainless Steel Hexnuts™ with PEEK ferrule fittings

Specifications

Type Injection Loop
Material Stainless Steel
Diameter (English) Inner 0.009 in.
Diameter (Metric) Inner 0.02 cm
Volume (Metric) 10 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.