Learn More
Eisco™ Trajectory Apparatus Kit - Model to demonstrate two-dimensional motion.
SCI_ED
Catalog No. S13024 Shop All Eisco Products
Change view
Click to view available options
38 x 43cm trajectory board
Catalog No.
S13024 38 x 43cm trajectory board
1 options
Catalog No. S13024 Supplier Eisco™ Supplier No. STRJ
Add to Cart
Add to Cart
Includes
38 X 43 cm trajectory board, metal target, two wooden supports, adjustable aluminum luncher with stop, steel ball, graph paper and pre-cut target paper
Model to demonstrate two-dimensional motion.
  • Steel ball decends metal ramp and strikes at opposing metal plate
  • Graph paper attached to back drop
  • Students plot different trajectories versus dropping height on graph paper
Specifications
Product Type Kit, Trajectory Apparatus
For Use With (Application) To demonstrate two-dimensional motion.
Includes 38 X 43 cm trajectory board, metal target, two wooden supports, adjustable aluminum luncher with stop, steel ball, graph paper and pre-cut target paper