Learn More
AdipoGen Tralomethrin
SDP
Supplier:  AdipoGen CDXT0116M500
Encompass_Preferred
Chemical. CAS 66841-25-6. Formula C22H19Br4NO3. MW 665.01. Synthetic. Potent pyrethroid insecticide. Mode of action is by modifying the gating kinetics of the sodium channels in neurons, increasing the length of time the channel remains open after a stimulus, thereby depolarizing the neuron for a longer period of time. This leads to uncontrolled spasming, paralysis and eventual death. It is effective against most members of class Arachnida as well as insects.
Catalog No. 50-255-9999
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.