Learn More
Enzo Life Sciences Trametinib (500mg). CAS: 871700-17-3
SDP
Supplier:  Enzo Life Sciences ENZCHM2390500
Encompass_Preferred
Trametinib is a highly specific and potent MEK1/2 inhibitor with IC50 of 0.92 nM/1.8 nM in cell-free assays. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (20 mg/mL). Long Term Storage: -20°C.
Catalog No. 50-201-3675
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.