Learn More
Apexbio Technology LLC Trametinib (GSK1120212), 871700-17-3, MFCD17215075, 50 mg
SDP
Supplier:  Apexbio Technology LLC A301850
Encompass_Preferred
MF: C26H23FIN5O4, MW: 615.39, Synonyms: Trametinib, GSK-1120212, GSK1120212, Mekinist, JTP74057, JTP-74057. Solubility: 15.38mg/mL in DMSO. Trametinib (also known as GSK1120212 or JTP 74057), originally identified as a p15 inductive compound, is a novel and potent allosteric inhibitor of MEK kinase, which exhibits ATP non-competitive inhibition against MEK1 and MEK2 kinase.
Catalog No. 50-101-3046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.