Learn More
Abcam Tramiprosate, Synthetic GABA Analog, Solid, 1 g
Description
- Molecular Weight: 139.18 Da
- CAS Number: 3687-18-1
- Form: Solid
- Activity: Binds soluble amyloid-β, exhibits antifibrillogenic activity in vitro and in vivo, and inhibits amyloid-β induced neuronal cytotoxicity
- Properties: Sulfated glycosaminoglycan (sGAG) mimetic, synthetic GABA analog, blood-brain barrier permeable, and orally active
- Composition: 100% 3-aminopropane-1-sulfonic acid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Amyloid-β |
| Substrate Properties | Chemical Substrate |
| Form | Solid |
| Shipping Condition | Ambient |
| Buffer | 100% 3-aminopropane-1-sulfonic acid |
| Content And Storage | Store at 4°C. |
| Formulation | 100% 3-aminopropane-1-sulfonic acid |
| Quantity | 1 g |
| For Use With (Application) | Amyloid-β research, Neurobiology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.