Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tramiprosate, Synthetic GABA Analog, Solid, 5 g

Catalog No. 502706889
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6889 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6889 Supplier Abcam Supplier No. AB1411165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic GABA analog with antifibrillogenic activity that binds soluble amyloid-β.

  • Molecular Weight: 139.18 Da
  • Form: Solid
  • Activity: Binds soluble amyloid-β, exhibits antifibrillogenic activity in vitro and in vivo, and inhibits amyloid-β induced neuronal cytotoxicity
  • Compound Type: Synthetic GABA analog and sulfated glycosaminoglycan (sGAG) mimetic
  • Properties: Blood-brain barrier permeable and orally active
  • Composition: 100% 3-aminopropane-1-sulfonic acid
  • CAS Number: 3687-18-1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 3-aminopropane-1-sulfonic acid
Form Solid
Content And Storage Store at 4°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Amyloid-beta
Formulation 100% 3-aminopropane-1-sulfonic acid
Quantity 5 g
For Use With (Application) Neuroscience-Amyloid-beta research, Neuronal cytotoxicity inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.