Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tran-35S-Label, No-Thaw Metabolic Labeling Reagent , MP Biomedical

Catalog No. MP01510094 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1.4 mci
14 mci
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP01510094 1.4 mci
MP015100914 14 mci
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP01510094 Supplier MP Biomedicals, Inc Supplier No. 0151009.4

Stabilized aqueous solution.

  • (L-Methionine, [35S]: L-Cysteine, [35S])
  • Sp. Act. >1000 Ci/mmol; >37.0 TBq/mmol
  • Net L-Methionine content 1 mCi, 5 mCi and 10 mCi, respectively
  • Metabolic labeling reagent derived from [35S]E. coli hydrolysate
  • Containing >70% L-methionine, [35S], ∼15% L-cysteine,[35S]
  • Stabilized aqueous solution
  • Concentration ∼10 mCi/ml; ∼370 MBq/ml
  • Packaged under nitrogen in the Versatainer™ System
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1.4 mci
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.