Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAN35S-LABEL, Metabolic Labeling Reagent, MP Biomedicals™

Catalog No. MP015100614 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
14mCi; 518MBq
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP015100614 14mCi; 518MBq
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP015100614 Supplier MP Biomedicals, Inc Supplier No. 015100614

Specifications

Physical Form Solution in 50 mM L-Lysine pH 7.4, containing 10 mM 2-mercaptoethanol
Chemical Name or Material TRAN35S-LABEL, Metabolic Labeling Reagent
Synonym L-Methionine, [35S]: L-Cysteine, [35S]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.