Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-trans-1,2-Cyclohexanedicarboxylic Anhydride 98.0+%, TCI America™
SDP

Catalog No. C165725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C165725G 25 g
C16575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C165725G Supplier TCI America Supplier No. C165725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14166-21-3
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
InChI Key MUTGBJKUEZFXGO-WDSKDSINSA-N
PubChem CID 12261141
IUPAC Name (3aS,7aS)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES C1CCC2C(C1)C(=O)OC2=O

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-trans-1,2-Cyclohexanedicarboxylic Anhydride
EINECSNumber (3)-2416
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.