Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,3-Cyclohexanediamine 97.0+%, TCI America™
SDP

Catalog No. C28021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28021G 1 g
C2802200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28021G Supplier TCI America Supplier No. C28021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26883-70-5
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
InChI Key GEQHKFFSPGPGLN-WDSKDSINSA-N
Synonym trans-1,3-Diaminocyclohexane
PubChem CID 6999482
IUPAC Name (1S,3S)-cyclohexane-1,3-diamine
SMILES C1CC(CC(C1)N)N

Specifications

Color Colorless
Boiling Point 198°C
Quantity 1 g
UN Number 2735
Formula Weight 114.19
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material trans-1,3-Cyclohexanediamine
EINECSNumber (3)-3871
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.