Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,3-Cyclohexanediol 98.0+%, TCI America™
SDP

Catalog No. C23231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23231G Supplier TCI America Supplier No. C23231G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5515-64-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00039458,MFCD16295186
InChI Key RLMGYIOTPQVQJR-UHFFFAOYNA-N
Synonym trans-1,3-Dihydroxycyclohexane
PubChem CID 6950744
IUPAC Name cyclohexane-1,3-diol
SMILES OC1CCCC(O)C1

Specifications

Melting Point 115°C
Color White-Yellow
Boiling Point 135°C
Quantity 1 g
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,3-Cyclohexanediol
EINECSNumber (3)-2310
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.