Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,4-Cyclohexanedicarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. C047525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C047525G 25 g
C0475100G 100 g
C0475500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C047525G Supplier TCI America Supplier No. C047525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 619-82-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00066203
InChI Key PXGZQGDTEZPERC-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane
PubChem CID 14106
IUPAC Name cyclohexane-1,4-dicarboxylic acid
SMILES C1CC(CCC1C(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 172.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,4-Cyclohexanedicarboxylic Acid
EINECSNumber (3)-2423&(3)-2446
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.