Learn More
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane, 97%
Description
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.
Solubility
Slightly Soluble (4.4 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Solubility Information | Slightly Soluble (4.4g/L) (25°C). |
| Quantity | 5 g |
| Formula Weight | 229.31 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.