Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-10-Hydroxy-2-decenoic Acid 97.0+%, TCI America™
SDP

Catalog No. H13371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13371G 1 g
H1337200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13371G Supplier TCI America Supplier No. H13371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14113-05-4
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00204506
InChI Key QHBZHVUGQROELI-SOFGYWHQSA-N
PubChem CID 5312738
ChEBI CHEBI:78668
IUPAC Name (2E)-10-hydroxydec-2-enoic acid
SMILES OCCCCCCC\C=C\C(O)=O

Specifications

Melting Point 66°C
Color White
Quantity 1 g
Formula Weight 186.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-10-Hydroxy-2-decenoic Acid
EINECSNumber (2)-1340
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.