Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam trans-11-Methyl Octadecenoate, Solid, >99% Purity, 100 mg

Catalog No. 502706060
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6060 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6060 Supplier Abcam Supplier No. AB143924100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

High-purity methyl ester of trans-11-octadecenoic acid for use in lipid studies and as an internal standard.

  • Purity: >99%
  • Molecular Weight: 296.5 Da
  • Form: Solid
  • Chemical Class: Methyl ester of trans-11-octadecenoic acid
  • Application: Use in lipid studies and as internal standard
  • Properties: Possible insulinotropic effects
  • Constituents: 100% Methyl (11E)-octadec-11-enoate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Methyl (11E)-octadec-11-enoate
Content And Storage Store at -20°C.
Form Solid
Quantity 100 mg
For Use With (Application) Lipid studies, internal standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.