Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2,3-Dimethylacrylic Acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
80-59-1
C5H8O2
100.12
MFCD00066864
UIERETOOQGIECD-ONEGZZNKSA-N
tiglic acid,tiglinic acid,cevadic acid,2-methyl-2-butenoic acid,trans-2-methylcrotonic acid,trans-2,3-dimethylacrylic acid,2-methylbut-2-enoic acid,e-2-methyl-2-butenoic acid,trans-2-methyl-2-butenoic acid,e-2,3-dimethylacrylic acid
125468
CHEBI:9592
(E)-2-methylbut-2-enoic acid
CC=C(C)C(=O)O
80-59-1
C5H8O2
100.12
MFCD00066864
UIERETOOQGIECD-ONEGZZNKSA-N
tiglic acid,tiglinic acid,cevadic acid,2-methyl-2-butenoic acid,trans-2-methylcrotonic acid,trans-2,3-dimethylacrylic acid,2-methylbut-2-enoic acid,e-2-methyl-2-butenoic acid,trans-2-methyl-2-butenoic acid,e-2,3-dimethylacrylic acid
125468
CHEBI:9592
(E)-2-methylbut-2-enoic acid
CC=C(C)C(=O)O
Color White to Beige
Melting Point 61.0°C to 65.0°C
Flash Point 95°C
Boiling Point 198.4°C
Infrared Spectrum Authentic
Specific Gravity 0.96
Quantity 25g
Packaging Glass bottle
Density 0.9600g/mL
Assay Percent Range 98%
Linear Formula CH3CH=C(CH3)CO2H
Beilstein 02, 430
Merck Index 15, 9589
Solubility Information Solubility in water: soluble in hot water. Other solubilities: 1g/10mL methanol,soluble in alcohol and ether
Formula Weight 100.12
Physical Form Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material trans-2, 3-Dimethylacrylic acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 201-295-