Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2,4-Dichlorocinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. D258825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D258825G 25 g
D25885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D258825G Supplier TCI America Supplier No. D258825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20595-45-3
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD00004373
InChI Key MEBWABJHRAYGFW-DUXPYHPUSA-N
Synonym 2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid
PubChem CID 688026
IUPAC Name (E)-3-(2,4-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O

Specifications

Melting Point 236°C
Color White
Quantity 25 g
Formula Weight 217.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,4-Dichlorocinnamic Acid
RTECSNumber GD8575000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.