Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2,5-Dimethylpiperazine 99.0+%, TCI America™
SDP

Catalog No. D076325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D076325G 25 g
D07635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D076325G Supplier TCI America Supplier No. D076325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2815-34-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00070516
InChI Key NSMWYRLQHIXVAP-OLQVQODUSA-N
Synonym trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine #
PubChem CID 220672
IUPAC Name (2R,5S)-2,5-dimethylpiperazine
SMILES CC1CNC(CN1)C

Specifications

Melting Point 116°C
Color White-Yellow
Boiling Point 162°C
Quantity 25 g
UN Number 2926
Formula Weight 114.19
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,5-Dimethylpiperazine
EINECSNumber (5)-0955
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.