Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Decenal 93.0+%, TCI America™
SDP

Catalog No. D164225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D164225ML 25 mL
D16425ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D164225ML Supplier TCI America Supplier No. D164225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3913-81-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00014679
InChI Key MMFCJPPRCYDLLZ-CMDGGOBGSA-N
Synonym trans-2-decenal, e-dec-2-enal, 2-decenal, decenal, e-2-decenal, 3-heptylacrolein, decenaldehyde, decylenic aldehyde, 2-decenal, 2e, 2-decenal, e
PubChem CID 5283345
IUPAC Name (2E)-dec-2-enal
SMILES CCCCCCC\C=C\C=O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Decenal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.