Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexene, 98+%

Catalog No. AC149900050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149900050 5 mL Glass bottle
AC149900250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC149900050 Supplier Thermo Scientific Chemicals Supplier No. 149900050
Add to Cart

CAS: 4050-45-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4050-45-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
PubChem CID 639661
IUPAC Name (E)-hex-2-ene
SMILES CCC\C=C\C

Specifications

Melting Point -98.5°C
Color Colorless
Density 0.6600g/mL
Boiling Point 68°C to 69°C
Flash Point -20°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3920 to 1.394
Linear Formula CH3CH2CH2CH=CHCH3
Quantity 5 mL
Beilstein 01, I, 89
Specific Gravity 0.66
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol
Formula Weight 84.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material trans-2-Hexene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-752-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.