Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methyl-2-butenal 95.0+%, TCI America™
SDP

Catalog No. T100325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100325ML 25 mL
T10035ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T100325ML Supplier TCI America Supplier No. T100325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 497-03-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006977
InChI Key ACWQBUSCFPJUPN-HWKANZROSA-N
Synonym tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
PubChem CID 5321950
IUPAC Name (E)-2-methylbut-2-enal
SMILES CC=C(C)C=O

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 mL
UN Number 1989
Formula Weight 84.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Methyl-2-butenal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.