Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methyl-2-butenoic acid, 97%, Thermo Scientific™

Catalog No. AAB2030803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2030803 1 g
AAB2030806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2030803 Supplier Thermo Scientific Chemicals Supplier No. B2030803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (2E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O

Specifications

Quantity 1 g
UN Number UN3261
Solubility Information Slightly soluble in water
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-butenoic acid
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.