Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Pentenoic acid, 97%

Catalog No. AC279370250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279370250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279370250 Supplier Thermo Scientific Chemicals Supplier No. 279370250
  • $116.50
    $99.65 / Each of 1

    Save $16.85

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13991-37-2 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13991-37-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00002704
InChI Key YIYBQIKDCADOSF-ONEGZZNKSA-N
Synonym trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
PubChem CID 638122
ChEBI CHEBI:38366
IUPAC Name (2E)-pent-2-enoic acid
SMILES CC\C=C\C(O)=O

Specifications

Melting Point 9.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 106.0°C (20.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C2H5CH=CHCO2H
Refractive Index 1.4522
Quantity 25 g
Beilstein 02, 426
Specific Gravity 0.99
Solubility Information Solubility in water: 60g/L (20°C)
Formula Weight 100.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-2-Pentenoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-791-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.