Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Undecenal 93.0+%, TCI America™
SDP

Catalog No. U004625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004625ML 25 mL
U00465ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U004625ML Supplier TCI America Supplier No. U004625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53448-07-0
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00014680
InChI Key PANBRUWVURLWGY-MDZDMXLPSA-N
Synonym trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30
PubChem CID 5283356
IUPAC Name (E)-undec-2-enal
SMILES CCCCCCCCC=CC=O

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 mL
Formula Weight 168.28
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Undecenal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.