Learn More
Sigma Organic Chemistry trans-4- 4-Morpholinyl | 1G | MFCD18071434
Supplier: Sigma Organic Chemistry CBR006751G

trans-4- 4-Morpholinyl , 1G
About this Item:
MDL #: MFCD18071434
Molecular Weight: 245.15
UNSPSC Code: 12352200
Chemical Formula: C8H18Cl2N2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.